Please login to your online account to display your discounted pricing

anti-ROR gamma/RORC/NR1F3, Clone: 4G419, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 4G419
Host Species Murine
Isotype κ
Immunogen A portion of amino acids 1-50 of human ROR gamma was used as the immunogen.
Antigen ROR gamma/RORC/NR1F3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224449

 
novus biologicals canada
NBP224449
Unlabeled 0.1mg Each for $329.00

NBP224449SS

 
novus biologicals canada
NBP224449SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 4G419
Host Species Murine
Isotype κ
Immunogen A portion of amino acids 1-50 of human ROR gamma was used as the immunogen.
Antigen ROR gamma/RORC/NR1F3
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human, 86% in Mouse and Rat, 80% in Bovine, and 73% in Chicken.
Gene Accession No. P51449
Gene ID (Entrez) 6097
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL