Please login to your online account to display your discounted pricing

anti-RIG-I, Clone: 2M6F10, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 2M6F10
Host Species Murine
Isotype κ
Immunogen A portion of amino acid 1-50 of human RIG-I was used as the immunogen. 31 PWFREEEVQYIQAEkNNkG 49
Antigen RIG-I
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227109

 
novus biologicals canada
NBP227109
Unlabeled 0.1mg Each for $329.00

NBP227109SS

 
novus biologicals canada
NBP227109SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 2M6F10
Host Species Murine
Isotype κ
Immunogen A portion of amino acid 1-50 of human RIG-I was used as the immunogen. 31 PWFREEEVQYIQAEkNNkG 49
Antigen RIG-I
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human, Chimp, Orangutan and Rhesus Monkey, 85% homologous in Pig, 79% homologous in Mouse and Horse and 73% homologous in Rat.
Gene Accession No. O95786
Gene ID (Entrez) 23586
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL