Please login to your online account to display your discounted pricing

anti-RIG-I, Clone: 2M6F10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227109SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227109SS

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Clone 2M6F10
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Immunogen A portion of amino acid 1-50 of human RIG-I was used as the immunogen. 31 PWFREEEVQYIQAEkNNkG 49
Host Species Mouse
Antigen RIG-I
Format Purified
Quantity 0.025mg
Regulatory Status RUO
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Gene Accession No. O95786
Gene ID (Entrez) 23586
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human, Chimp, Orangutan and Rhesus Monkey, 85% homologous in Pig, 79% homologous in Mouse and Horse and 73% homologous in Rat.