Please login to your online account to display your discounted pricing

anti-Rhodopsin, Clone: A531, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone A531
Host Species Murine
Isotype κ
Immunogen Whole purified bovine Rhodopsin [UniProt No. P02699]
Antigen Rhodopsin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225159

 
novus biologicals canada
NBP225159
Unlabeled 0.1mL Each for $309.05

NBP225159SS

 
novus biologicals canada
NBP225159SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone A531
Host Species Murine
Isotype κ
Immunogen Whole purified bovine Rhodopsin [UniProt No. P02699]
Antigen Rhodopsin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human, Bovine, Mouse, Porcine and Rat. Other species have not been tested.
Gene Accession No. P08100
Gene ID (Entrez) 6010
Formulation PBS
Concentration 1mg/mL