Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Clone: 112A1021, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227416

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227416

  • / Each
 

Description & Specifications

Specifications

Immunogen This antibody was raised against a synthetic peptide corresponding to amino acids 526-539 of human NF-kB (p65) protein.
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mg
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Mouse
Antigen RelA/NFkB p65
Test Specificity Cross reacts with Human, Mouse and Rat. Predicted to react with Cow, Gorilla, New World Monkey and Pig.
Gene Accession No. Q969V5
Concentration 0.5mg/mL
Primary or Secondary Primary
Formulation 0.1mg of antibody in 0.2mL sterile-filtered PBS.
Gene ID (Entrez) 5970
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Isotype κ
Format Purified
Clone 112A1021