Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Clone: 112A1021, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056712

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056712

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Immunogen This antibody was raised against a synthetic peptide corresponding to amino acids 526-539 of human NF-kB (p65) protein.
Gene ID (Entrez) 5970
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. Q969V5
Quantity 0.1mg
Clone 112A1021
Concentration 0.5mg/mL
Regulatory Status RUO
Antigen RelA/NFkB p65
Isotype IgG1, κ
Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Cross reacts with Human, Mouse and Rat. Predicted to react with Cow, Gorilla, New World Monkey and Pig.
Conjugate Unlabeled