Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Clone: 112A1021, Novus Biologicals™

$192.00 - $406.00

Specifications

Clone 112A1021
Host Species Mouse
Isotype κ
Immunogen This antibody was raised against a synthetic peptide corresponding to amino acids 526-539 of human NF-kB (p65) protein.
Antigen RelA/NFkB p65
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227416SS

 
novus biologicals canada
NBP227416SS
Unlabeled 0.025mg Each for $192.00

NBP227416

 
novus biologicals canada
NBP227416
Unlabeled 0.1mg Each for $406.00
Description & Specifications

Specifications

Clone 112A1021
Host Species Mouse
Isotype κ
Immunogen This antibody was raised against a synthetic peptide corresponding to amino acids 526-539 of human NF-kB (p65) protein.
Antigen RelA/NFkB p65
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Cross reacts with Human, Mouse and Rat. Predicted to react with Cow, Gorilla, New World Monkey and Pig.
Gene Accession No. Q969V5
Gene ID (Entrez) 5970
Formulation 0.1mg of antibody in 0.2mL sterile-filtered PBS.
Concentration 0.5mg/mL