Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Clone: 112A1021, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227416SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227416SS

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen This antibody was raised against a synthetic peptide corresponding to amino acids 526-539 of human NF-kB (p65) protein.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype κ
Quantity 0.025mg
Regulatory Status RUO
Format Purified
Clone 112A1021
Formulation 0.1mg of antibody in 0.2mL sterile-filtered PBS.
Monoclonal or Polyclonal Monoclonal
Test Specificity Cross reacts with Human, Mouse and Rat. Predicted to react with Cow, Gorilla, New World Monkey and Pig.
Primary or Secondary Primary
Gene ID (Entrez) 5970
Concentration 0.5mg/mL
Conjugate Unlabeled
Gene Accession No. Q969V5
Antigen RelA/NFkB p65
Host Species Mouse