Please login to your online account to display your discounted pricing

anti-Recoverin, Clone: 6A55CD6, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300280SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300280SS

  • / Each
 

Description & Specifications

Specifications

Quantity 0.025mL
Antigen Recoverin
Cross Reactivity Human
Regulatory Status RUO
Formulation Tris-glycine, 150mM NaCl
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P35243
Primary or Secondary Primary
Conjugate Unlabeled
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene ID (Entrez) 5957
Isotype IgG2a, κ
Immunogen Full length recombinant human Recoverin. [UniProt No. P35243]
Host Species Mouse
Clone 6A55CD6
Format Purified
Concentration 4.5mg/mL