Please login to your online account to display your discounted pricing

anti-Recoverin, Clone: 6A55CD6, Novus Biologicals™

$139.00 - $139.00

Specifications

Clone 6A55CD6
Host Species Murine
Isotype IgG2a, κ
Immunogen Full length recombinant human Recoverin. [UniProt No. P35243]
Antigen Recoverin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300280SS

 
novus biologicals canada
NB300280SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 6A55CD6
Host Species Murine
Isotype IgG2a, κ
Immunogen Full length recombinant human Recoverin. [UniProt No. P35243]
Antigen Recoverin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. P35243
Gene ID (Entrez) 5957
Formulation Tris-glycine, 150mM NaCl
Concentration 4.5mg/mL