Please login to your online account to display your discounted pricing

anti-Recoverin, Clone: 6A55CD6, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300280

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300280

  • / Each
 

Description & Specifications

Specifications

Antigen Recoverin
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.1mL
Immunogen Full length recombinant human Recoverin. [UniProt No. P35243]
Gene ID (Entrez) 5957
Gene Accession No. P35243
Primary or Secondary Primary
Concentration 4.5mg/mL
Conjugate Unlabeled
Format Purified
Host Species Mouse
Isotype IgG2a, κ
Regulatory Status RUO
Cross Reactivity Human
Formulation Tris-glycine, 150mM NaCl
Monoclonal or Polyclonal Monoclonal
Clone 6A55CD6