Please login to your online account to display your discounted pricing

anti-RANK/TNFRSF11A, Clone: 9A725, Novus Biologicals™

$139.00 - $362.00

Specifications

Clone 9A725
Host Species Murine
Isotype IgG1, κ
Immunogen A fusion protein containing amino acid residues 326-616 of human RANK was used as immunogen. This antibody recognizes an epitope located between amino acid residues 330-427 of human RANK (NP_003830).
Antigen RANK/TNFRSF11A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056508T

 
novus biologicals canada
NB10056508SS
Unlabeled 0.025mg Each for $139.00

NB10056508

 
novus biologicals canada
NB10056508
Unlabeled 0.1mg Each for $362.00
Description & Specifications

Specifications

Clone 9A725
Host Species Murine
Isotype IgG1, κ
Immunogen A fusion protein containing amino acid residues 326-616 of human RANK was used as immunogen. This antibody recognizes an epitope located between amino acid residues 330-427 of human RANK (NP_003830).
Antigen RANK/TNFRSF11A
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity In RAW cells, a 97 kDa is observed. Cross reacts with Human and Mouse. Not yet tested in other species.
Gene Accession No. Q9Y6Q6
Gene ID (Entrez) 8792
Formulation PBS containing 0.5% BSA
Concentration 0.5mg/mL