Please login to your online account to display your discounted pricing

anti-RAD18, Clone: 79B1048.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056523

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056523

  • / Each
 

Description & Specifications

Specifications

Antigen RAD18
Applications Western Blot
Clone 79B1048.1
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Host Species Murine
Immunogen The antibody was raised against a synthetic peptide corresponding to the amino acids 402-414 of human Rad18.
Isotype IgG1
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 56852
Test Specificity In HL60, a 54 kDa band should be detected.
Target Species Human