Please login to your online account to display your discounted pricing

anti-Prolactin R, Clone: PRLR742, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232899

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232899

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Clone PRLR742
Antigen Prolactin R
Quantity 0.5mL
Cross Reactivity Human
Gene Accession No. P16471
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 5618
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Regulatory Status RUO
Immunogen Recombinant human prolactin receptor