Please login to your online account to display your discounted pricing

anti-Prolactin R, Clone: PRLR742, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone PRLR742
Host Species Mouse
Isotype κ
Immunogen Recombinant human prolactin receptor
Antigen Prolactin R
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328991M

 
novus biologicals canada
NBP2328991ML
Unlabeled 1mL Each for $429.00

NBP232899

 
novus biologicals canada
NBP232899
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone PRLR742
Host Species Mouse
Isotype κ
Immunogen Recombinant human prolactin receptor
Antigen Prolactin R
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P16471
Gene ID (Entrez) 5618
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL