Please login to your online account to display your discounted pricing

anti-Prolactin R, Clone: PRLR742, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328991ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328991M

  • / Each
 

Description & Specifications

Specifications

Immunogen Recombinant human prolactin receptor
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone PRLR742
Isotype κ
Host Species Mouse
Quantity 1mL
Gene ID (Entrez) 5618
Gene Accession No. P16471
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Cross Reactivity Human
Antigen Prolactin R
Regulatory Status RUO
Format Purified