Please login to your online account to display your discounted pricing

anti-Prolactin R, Clone: B6.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232991

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232991

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Regulatory Status RUO
Quantity 0.5mL
Cross Reactivity Human
Immunogen Semi-purified human prolactin receptor
Clone B6.2
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA.
Conjugate Unlabeled
Gene Accession No. P16471
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 5618
Antigen Prolactin R
Format Purified
Host Species Mouse