Please login to your online account to display your discounted pricing

anti-Prolactin R, Clone: B6.2, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone B6.2
Host Species Mouse
Isotype κ
Immunogen Semi-purified human prolactin receptor
Antigen Prolactin R
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232991

 
novus biologicals canada
NBP232991
Unlabeled 0.5mL Each for $321.00

NBP2329911M

 
novus biologicals canada
NBP2329911ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone B6.2
Host Species Mouse
Isotype κ
Immunogen Semi-purified human prolactin receptor
Antigen Prolactin R
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P16471
Gene ID (Entrez) 5618
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL