Please login to your online account to display your discounted pricing

anti-Procollagen Type 2A, Clone: 003-02, Novus Biologicals™

$142.00 - $406.15

Specifications

Clone 003-02
Host Species Mouse
Isotype IgG1, κ
Immunogen Recombinant expressed PIIANP
Antigen Procollagen Type 2A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP105169

 
novus biologicals canada
NBP105169
Unlabeled 0.1mg Each for $406.15

NBP105169SS

 
novus biologicals canada
NBP105169SS
Unlabeled 0.02mg Each for $142.00
Description & Specifications

Specifications

Clone 003-02
Host Species Mouse
Isotype IgG1, κ
Immunogen Recombinant expressed PIIANP
Antigen Procollagen Type 2A
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Procollagen type IIA N-terminal propeptide.
Gene Accession No. P08123
Gene ID (Entrez) 1278
Formulation 0.01M phosphate buffer, pH 7.4, with 0.5M NaCl
Concentration 1.0mg/mL