Please login to your online account to display your discounted pricing

anti-Procollagen Type 2A, Clone: 003-02, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP105169SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP105169SS

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Cross Reactivity Canine
Cross Reactivity Human
Isotype IgG1, κ
Host Species Mouse
Antigen Procollagen Type 2A
Formulation 0.01M phosphate buffer, pH 7.4, with 0.5M NaCl
Format Purified
Monoclonal or Polyclonal Monoclonal
Clone 003-02
Test Specificity Procollagen type IIA N-terminal propeptide.
Quantity 0.02mg
Concentration 1.0mg/mL
Immunogen Recombinant expressed PIIANP
Gene ID (Entrez) 1278
Conjugate Unlabeled
Gene Accession No. P08123
Primary or Secondary Primary