Please login to your online account to display your discounted pricing

anti-Presenilin-2, Clone: 198C679.2., Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056677

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056677

  • / Each
 

Description & Specifications

Specifications

Quantity 0.2mL
Regulatory Status RUO
Format Supernatant
Host Species Mouse
Applications Western Blotting
Immunogen This antibody was generated by immunizing mice with a synthetic peptide of human Presenilin-2 (PS-2).
Monoclonal or Polyclonal Monoclonal
Test Specificity This tested negative in mouse NIH 3T3 cells.
Antigen Presenilin-2
Isotype IgM
Clone 198C679.2.
Gene Accession No. P49810
Gene ID (Entrez) 5664
Conjugate Unlabeled
Cross Reactivity Human
Formulation Tissue culture supernatant
Primary or Secondary Primary