Please login to your online account to display your discounted pricing

anti-Presenilin-2, Clone: 198C679.2., Novus Biologicals™

$139.00 - $259.02

Specifications

Clone 198C679.2.
Host Species Murine
Isotype IgM
Immunogen This antibody was generated by immunizing mice with a synthetic peptide of human Presenilin-2 (PS-2).
Antigen Presenilin-2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056677T

 
novus biologicals canada
NB10056677SS
Unlabeled 0.05mL Each for $139.00

NB10056677

 
novus biologicals canada
NB10056677
Unlabeled 0.2mL Each for $259.02
Description & Specifications

Specifications

Clone 198C679.2.
Host Species Murine
Isotype IgM
Immunogen This antibody was generated by immunizing mice with a synthetic peptide of human Presenilin-2 (PS-2).
Antigen Presenilin-2
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This tested negative in mouse NIH 3T3 cells.
Gene Accession No. P49810
Gene ID (Entrez) 5664
Formulation Tissue culture supernatant