Please login to your online account to display your discounted pricing

anti-Presenilin-2, Clone: 198C679.2., Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056677SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056677T

  • / Each
 

Description & Specifications

Specifications

Antigen Presenilin-2
Applications Western Blot
Clone 198C679.2.
Conjugate Unlabeled
Cross Reactivity Human
Format Supernatant
Formulation Tissue culture supernatant
Gene Accession No. P49810
Host Species Murine
Immunogen This antibody was generated by immunizing mice with a synthetic peptide of human Presenilin-2 (PS-2).
Isotype IgM
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5664
Test Specificity This tested negative in mouse NIH 3T3 cells.
Target Species Human