Please login to your online account to display your discounted pricing

anti-PPP2R5C, Clone: TQ11-1G6, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100501

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100501

  • / Each
 

Description & Specifications

Specifications

Immunogen Human PPP2R5C [UniProt No. Q13362]
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 5527
Test Specificity This is specific for PP2A (B56 gamma 3 subunit) protein. This does not recognize the B56 gamma 1 isoform of PP2A.
Gene Accession No. Q13362
Host Species Mouse
Isotype IgG1, κ
Regulatory Status RUO
Conjugate Unlabeled
Antigen PPP2R5C
Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Clone TQ11-1G6
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS
Primary or Secondary Primary
Concentration 2mg/mL