Please login to your online account to display your discounted pricing

anti-PPP2R5C, Clone: TQ11-1G6, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone TQ11-1G6
Host Species Mouse
Isotype IgG1, κ
Immunogen Human PPP2R5C [UniProt No. Q13362]
Antigen PPP2R5C
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100501

 
novus biologicals canada
NB100501
Unlabeled 0.1mL Each for $309.05

NB100501SS

 
novus biologicals canada
NB100501SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone TQ11-1G6
Host Species Mouse
Isotype IgG1, κ
Immunogen Human PPP2R5C [UniProt No. Q13362]
Antigen PPP2R5C
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This is specific for PP2A (B56 gamma 3 subunit) protein. This does not recognize the B56 gamma 1 isoform of PP2A.
Gene Accession No. Q13362
Gene ID (Entrez) 5527
Formulation PBS
Concentration 2mg/mL