Please login to your online account to display your discounted pricing

anti-PON1, Clone: 4G8D3, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 4G8D3
Host Species Murine
Isotype IgG1
Immunogen Partial recombinant human PON1 (between residues 1-200) expressed in &italic_begin;E. coli&italic_end; [Uniprot P27169]
Antigen PON1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223610SS

 
novus biologicals canada
NBP223610SS
Unlabeled 0.025mL Each for $109.00

NBP223610

 
novus biologicals canada
NBP223610
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 4G8D3
Host Species Murine
Isotype IgG1
Immunogen Partial recombinant human PON1 (between residues 1-200) expressed in &italic_begin;E. coli&italic_end; [Uniprot P27169]
Antigen PON1
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Western Blot
Gene Accession No. P27169
Gene ID (Entrez) 5444
Formulation PBS, 0.5% protein stabilizer
Concentration 1.0mg/mL