Please login to your online account to display your discounted pricing

anti-PMEL17/SILV, Clone: HMB45, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone HMB45
Host Species Mouse
Isotype κ
Immunogen An extract of pigmented melanoma metastases from lymph nodes was used as the immunogen for the melanoma HMB45 antibody.
Antigen PMEL17/SILV
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2294181M

 
novus biologicals canada
NBP2294181ML
Unlabeled 1mL Each for $429.00

NBP229418

 
novus biologicals canada
NBP229418
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone HMB45
Host Species Mouse
Isotype κ
Immunogen An extract of pigmented melanoma metastases from lymph nodes was used as the immunogen for the melanoma HMB45 antibody.
Antigen PMEL17/SILV
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity By immunohistochemistry, it specifically recognizes a protein in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. Metastatic amelanotic melanoma can often be confused with a variety of poorly differentiated carcinomas, large cell lymphomas, and sarcomas using H & E stains alone. It is also difficult to differentiate melanoma from spindle cell carcinomas and various types of mesenchymal neoplasms. Anti-HMB-45 stains fetal and neonatal melanocytes, junctional and blue nevus cells, and malignant melanoma. This MAb also stains Angiomyolipoma (PEComa). Canine (Negative), Feline (Negative).
Gene Accession No. P40967
Gene ID (Entrez) 6490
Formulation Culture Medium