Please login to your online account to display your discounted pricing

anti-PKC alpha, Clone: MC5, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone MC5
Host Species Murine
Isotype IgG2a
Immunogen Purified bovine brain Protein Kinase C
Antigen PKC alpha
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600201

 
novus biologicals canada
NB600201
Unlabeled 0.1mL Each for $329.00

NB600201SS

 
novus biologicals canada
NB600201SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone MC5
Host Species Murine
Isotype IgG2a
Immunogen Purified bovine brain Protein Kinase C
Antigen PKC alpha
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Radioimmunoassay
Applications Western Blot
Test Specificity This antibody is capable of detecting PKC beta as well.
Gene Accession No. P17252
Gene ID (Entrez) 5578
Formulation Tris-glycine, 150mM NaCl
Concentration 1.0mg/mL