Please login to your online account to display your discounted pricing

anti-PGLYRP4/PGRP-I beta, Clone: 186C426, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056721SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056721T

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 95-110 of human PGRP-1 beta.
Gene ID (Entrez) 57115
Concentration 0.5mg/mL
Antigen PGLYRP4/PGRP-I beta
Format Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Clone 186C426
Host Species Mouse
Quantity 0.025mg
Test Specificity PGRP-1b (186C426).
Conjugate Unlabeled
Cross Reactivity Human
Formulation PBS containing 0.0.5% BSA
Gene Accession No. Q96LB8
Primary or Secondary Primary