Please login to your online account to display your discounted pricing

anti-PGLYRP3/PGRPIA, Clone: 187C434, Novus Biologicals™

$139.00 - $259.02

Specifications

Clone 187C434
Host Species Murine
Isotype IgM
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 178-193 of human PGRP-1a.
Antigen PGLYRP3/PGRPIA
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056729T

 
novus biologicals canada
NB10056729SS
Unlabeled 0.05mL Each for $139.00

NB10056729

 
novus biologicals canada
NB10056729
Unlabeled 0.2mL Each for $259.02
Description & Specifications

Specifications

Clone 187C434
Host Species Murine
Isotype IgM
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 178-193 of human PGRP-1a.
Antigen PGLYRP3/PGRPIA
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity PGRP-1a (187C434).
Gene Accession No. Q96LB9
Gene ID (Entrez) 114771