Please login to your online account to display your discounted pricing

anti-PGLYRP3/PGRPIA, Clone: 187C434, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056729SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056729T

  • / Each
 

Description & Specifications

Specifications

Quantity 0.05mL
Format Ascites
Isotype IgM
Monoclonal or Polyclonal Monoclonal
Gene Accession No. Q96LB9
Conjugate Unlabeled
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 178-193 of human PGRP-1a.
Host Species Mouse
Cross Reactivity Human
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Antigen PGLYRP3/PGRPIA
Regulatory Status RUO
Clone 187C434
Test Specificity PGRP-1a (187C434).
Gene ID (Entrez) 114771
Primary or Secondary Primary