Please login to your online account to display your discounted pricing

anti-PGLYRP3/PGRPIA, Clone: 187C434, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056729

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056729

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Host Species Mouse
Antigen PGLYRP3/PGRPIA
Primary or Secondary Primary
Isotype IgM
Gene Accession No. Q96LB9
Quantity 0.2mL
Monoclonal or Polyclonal Monoclonal
Test Specificity PGRP-1a (187C434).
Format Ascites
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 178-193 of human PGRP-1a.
Conjugate Unlabeled
Cross Reactivity Human
Gene ID (Entrez) 114771
Clone 187C434