Please login to your online account to display your discounted pricing

anti-PGLYRP1/PGRP-S, Clone: 188C424, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056719

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056719

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8993
Isotype IgG3, κ
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Clone 188C424
Antigen PGLYRP1/PGRP-S
Quantity 0.1mg
Primary or Secondary Primary
Concentration 0.5mg/mL
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 165-180 (YVLKGHRDVQRTLSPG) of human PGRP-S (NP_005082).
Gene Accession No. O75594
Formulation PBS containing 0.05% BSA