Please login to your online account to display your discounted pricing

anti-PGLYRP1/PGRP-S, Clone: 188C424, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 188C424
Host Species Murine
Isotype IgG3, κ
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 165-180 (YVLKGHRDVQRTLSPG) of human PGRP-S (NP_005082).
Antigen PGLYRP1/PGRP-S
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056719

 
novus biologicals canada
NB10056719
Unlabeled 0.1mg Each for $329.00

NB10056719T

 
novus biologicals canada
NB10056719SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 188C424
Host Species Murine
Isotype IgG3, κ
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 165-180 (YVLKGHRDVQRTLSPG) of human PGRP-S (NP_005082).
Antigen PGLYRP1/PGRP-S
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blot
Gene Accession No. O75594
Gene ID (Entrez) 8993
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL