Please login to your online account to display your discounted pricing

anti-PGLYRP1/PGRP-S, Clone: 188C424, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056719SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056719T

  • / Each
 

Description & Specifications

Specifications

Antigen PGLYRP1/PGRP-S
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blot
Clone 188C424
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. O75594
Host Species Murine
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 165-180 (YVLKGHRDVQRTLSPG) of human PGRP-S (NP_005082).
Isotype IgG3, κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8993
Target Species Human
Target Species Murine