Please login to your online account to display your discounted pricing

anti-Peripherin, Clone: 7C5, Novus Biologicals™


Manufacturer: novus biologicals canada  NB30022002ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB30022002M

  • / Each
 

Description & Specifications

Specifications

Immunogen Recombinant rat Peripherin purified from &italic_begin;E. coli&italic_end;. [UniProt No. P21807]
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.2mL
Regulatory Status RUO
Clone 7C5
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Rat
Format Supernatant
Isotype IgG1, κ
Gene ID (Entrez) 5630
Host Species Mouse
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Chicken. Expected to cross-react with all mammals due to sequence homology.
Antigen Peripherin