Please login to your online account to display your discounted pricing

anti-Pax7, Clone: PAX7/497, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232894

 View all options for this product


For Research Use Only

Catalog No. NBP232894


Description & Specifications


Antigen Pax7
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone PAX7/497
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Zebrafish
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P23759
Host Species Mouse
Immunogen Recombinant human PAX7 protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5081