Please login to your online account to display your discounted pricing

anti-Pax7, Clone: PAX7/497, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone PAX7/497
Host Species Mouse
Isotype κ
Immunogen Recombinant human PAX7 protein
Antigen Pax7
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328941M

 
novus biologicals canada
NBP2328941ML
Unlabeled 1mL Each for $429.00

NBP232894

 
novus biologicals canada
NBP232894
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone PAX7/497
Host Species Mouse
Isotype κ
Immunogen Recombinant human PAX7 protein
Antigen Pax7
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P23759
Gene ID (Entrez) 5081
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL