Please login to your online account to display your discounted pricing

anti-Pax7, Clone: PAX7/497, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328941ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328941M

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Zebrafish
Antigen Pax7
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human PAX7 protein
Host Species Mouse
Clone PAX7/497
Conjugate Unlabeled
Primary or Secondary Primary
Concentration 0.2mg/mL
Gene Accession No. P23759
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 5081
Quantity 1mL
Regulatory Status RUO