Please login to your online account to display your discounted pricing

anti-Pax6, Clone: PAX6/496, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232893

 View all options for this product


For Research Use Only

Catalog No. NBP232893


Description & Specifications


Antigen Pax6
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone PAX6/496
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P26367
Host Species Mouse
Immunogen Recombinant human PAX6 protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5080