Please login to your online account to display your discounted pricing

anti-Pax6, Clone: PAX6/496, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone PAX6/496
Host Species Mouse
Isotype κ
Immunogen Recombinant human PAX6 protein
Antigen Pax6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232893

 
novus biologicals canada
NBP232893
Unlabeled 0.5mL Each for $321.00

NBP2328931M

 
novus biologicals canada
NBP2328931ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone PAX6/496
Host Species Mouse
Isotype κ
Immunogen Recombinant human PAX6 protein
Antigen Pax6
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P26367
Gene ID (Entrez) 5080
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL