Please login to your online account to display your discounted pricing

anti-Pax6, Clone: PAX6/496, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328931ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328931M

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 1mL
Immunogen Recombinant human PAX6 protein
Antigen Pax6
Concentration 0.2mg/mL
Gene Accession No. P26367
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 5080
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Primary or Secondary Primary
Regulatory Status RUO
Host Species Mouse
Format Purified
Clone PAX6/496