Please login to your online account to display your discounted pricing

anti-PARP, Clone: 194C1439, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056599SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056599T

  • / Each
 

Description & Specifications

Specifications

Antigen PARP
Applications Flow Cytometry
Applications Western Blot
Clone 194C1439
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. P09874
Host Species Murine
Immunogen This antibody was developed by immunizing mice with a synthetic peptide containing amino acids near 214/215-cleavage site of human PARP.
Isotype IgG2b, κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 142
Test Specificity This monoclonal antibody specifically detects a 89-kDa cleavage fragment containing the catalytic and auto modification domain.
Target Species Human