Please login to your online account to display your discounted pricing

anti-p73, Clone: 5B789, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056675

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056675

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Isotype IgG1, κ
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Applications Western Blotting
Immunogen This antibody was raised against full-length human p73. IMG-260 reacts with an epitope that is located on the C-terminal region of human p73 only. It does not cross react with mouse p73 or with p53.
Gene ID (Entrez) 7161
Clone 5B789
Gene Accession No. O15350
Concentration 0.5mg/mL
Regulatory Status RUO
Cross Reactivity Human
Antigen p73
Conjugate Unlabeled
Test Specificity Caspase-cleaved p73 fragments, including 55 kDa and 45 kDa, have been described (Sayan et al, 2008). The ability of a given antibody to recognize a p73 fragment will depend on whether or not the antibody epitope is contained in the fragment. It does not cross react with mouse p73 or with p53.
Format Purified
Formulation PBS containing 0.05% BSA
Primary or Secondary Primary