Please login to your online account to display your discounted pricing

anti-p73, Clone: 5B789, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 5B789
Host Species Mouse
Isotype IgG1, κ
Immunogen This antibody was raised against full-length human p73. IMG-260 reacts with an epitope that is located on the C-terminal region of human p73 only. It does not cross react with mouse p73 or with p53.
Antigen p73
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056675

 
novus biologicals canada
NB10056675
Unlabeled 0.1mg Each for $329.00

NB10056675T

 
novus biologicals canada
NB10056675SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 5B789
Host Species Mouse
Isotype IgG1, κ
Immunogen This antibody was raised against full-length human p73. IMG-260 reacts with an epitope that is located on the C-terminal region of human p73 only. It does not cross react with mouse p73 or with p53.
Antigen p73
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Caspase-cleaved p73 fragments, including 55 kDa and 45 kDa, have been described (Sayan et al, 2008). The ability of a given antibody to recognize a p73 fragment will depend on whether or not the antibody epitope is contained in the fragment. It does not cross react with mouse p73 or with p53.
Gene Accession No. O15350
Gene ID (Entrez) 7161
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL