Please login to your online account to display your discounted pricing

anti-p73, Clone: 5B789, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056675SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056675T

  • / Each
 

Description & Specifications

Specifications

Antigen p73
Applications Western Blot
Clone 5B789
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. O15350
Host Species Murine
Immunogen This antibody was raised against full-length human p73. IMG-260 reacts with an epitope that is located on the C-terminal region of human p73 only. It does not cross react with mouse p73 or with p53.
Isotype IgG1, κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7161
Test Specificity Caspase-cleaved p73 fragments, including 55 kDa and 45 kDa, have been described (Sayan et al, 2008). The ability of a given antibody to recognize a p73 fragment will depend on whether or not the antibody epitope is contained in the fragment. It does not cross react with mouse p73 or with p53.
Target Species Human