Please login to your online account to display your discounted pricing

anti-p73, Clone: 5B1288, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056674SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056674T

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1, κ
Host Species Mouse
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Gene Accession No. O15350
Primary or Secondary Primary
Concentration 0.5mg/mL
Conjugate Unlabeled
Gene ID (Entrez) 7161
Quantity 0.025mg
Immunogen This antibody was raised against full-length human p73. The epitope is thought to lie around the center of the molecule (NP_005418).
Test Specificity It reacts with alpha, beta, gamma and delta isoforms of mouse and human p73 as well as the dominant negative p73 (Costanzo, et al, 2002). Because of its reactivity pattern it may be regarded as anti-pan p73. Immunoprecipitation details can be found in Sayan et al (2005). NB100-56674 has been shown to recongize all the known alternative splicing variants of human and mouse p73 (Costanzo et al, 2002). The antibody does not cross react with p53.
Format Purified
Antigen p73
Clone 5B1288
Regulatory Status RUO
Formulation PBS containing 0.05% BSA