Please login to your online account to display your discounted pricing

anti-p73, Clone: 5B1288, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056674

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056674

  • / Each
 

Description & Specifications

Specifications

Antigen p73
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Clone 5B1288
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. O15350
Host Species Murine
Immunogen This antibody was raised against full-length human p73. The epitope is thought to lie around the center of the molecule (NP_005418).
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7161
Test Specificity It reacts with alpha, beta, gamma and delta isoforms of mouse and human p73 as well as the dominant negative p73 (Costanzo, et al, 2002). Because of its reactivity pattern it may be regarded as anti-pan p73. Immunoprecipitation details can be found in Sayan et al (2005). NB100-56674 has been shown to recongize all the known alternative splicing variants of human and mouse p73 (Costanzo et al, 2002). The antibody does not cross react with p53.
Target Species Human
Target Species Murine