Please login to your online account to display your discounted pricing

anti-p62/SQSTM1, Clone: 5H7E2, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 5H7E2
Host Species Murine
Isotype IgG1
Immunogen Partial recombinant human p62/SQSTM1 (between residues 100-400) expressed in &italic_begin;E. coli&italic_end; [UniProt Q13501]
Antigen p62/SQSTM1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223490SS

 
novus biologicals canada
NBP223490SS
Unlabeled 0.025mL Each for $139.00

NBP223490

 
novus biologicals canada
NBP223490
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 5H7E2
Host Species Murine
Isotype IgG1
Immunogen Partial recombinant human p62/SQSTM1 (between residues 100-400) expressed in &italic_begin;E. coli&italic_end; [UniProt Q13501]
Antigen p62/SQSTM1
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q13501
Gene ID (Entrez) 8878
Formulation PBS, 0.5% protein stabilizer
Concentration 1.0mg/mL