Please login to your online account to display your discounted pricing

anti-p62/SQSTM1, Clone: 5H7E2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223490SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223490SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene Accession No. Q13501
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8878
Concentration 1.0mg/mL
Cross Reactivity Human
Cross Reactivity Primate
Format Precipitation
Immunogen Partial recombinant human p62/SQSTM1 (between residues 100-400) expressed in &italic_begin;E. coli&italic_end; [UniProt Q13501]
Isotype IgG1
Quantity 0.025mL
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Antigen p62/SQSTM1
Host Species Mouse
Formulation PBS, 0.5% protein stabilizer
Clone 5H7E2
Conjugate Unlabeled